Orders LinkContactUsLinkComponent
Stopcocks, Rotaflo®
Stopcocks, Rotaflo®
3 mm, 8 mm
  201-8222
 :   SciLabware
Stopcocks, Rotaflo®
  201-8222
Stopcocks, Rotaflo®
Stopcocks, Rotaflo®
3 mm, 8 mm
265,00 €
1 * 5 items
PYREX®, borosilicate glass with PTFE needle valve.
  • Lubricant is not required (no seizure, no contamination with grease)
  • Interchangeable elements
  • Precise flow control
These chemically inert stopcocks can be used at temperatures up to +200 °C. Two different models are available: general purpose (vacuum rating up to 10-4 mm Hg) and high performance (vacuum rating up to 10-9 mm Hg).
  • Hole Ø:
    3 mm
  • Seq:
    01
  • For tube I-Ø:
    8 mm
Accessories
  •  
    Description
    For
  • 229-0230
    Replacement key, bore Ø 6 mm
    High performance Rotaflo® stopcocks
    1 * 2 items - 113,00 €
  •  
    Description
    For
  • 229-0231
    Replacement key, bore Ø 6 mm
    General purpose Rotaflo® stopcocks
    1 * 5 items - 156,00 €
  •  
    Description
    For
  • BIBBLR6
    Replacement locking ring for key with bore 6 mm
    Rotaflo® stopcocks
    1 * 5 items - 37,20 €
  •  
    Description
    For
  • 229-0226
    Replacement key, bore Ø 10 mm
    High performance Rotaflo® stopcocks
    1 * 1 items - 72,30 €
  •  
    Description
    For
  • 229-0229
    Replacement key, bore Ø 3 mm
    High performance Rotaflo® stopcocks
    1 * 2 items - 80,60 €
  •  
    Description
    For
  • BIBBLR3
    Replacement locking ring for key with bore 3 mm
    Rotaflo® stopcocks
    1 * 5 items - 25,40 €
  •  
    Description
    For
  • 229-0234
    Replacement key, bore Ø 10 mm
    General purpose Rotaflo® stopcocks
    1 * 5 items - 242,00 €
  •  
    Description
    For
  • BIBBGP3K
    Replacement key, bore Ø 3 mm
    General purpose Rotaflo® stopcocks
    1 * 5 items - 99,40 €
  •  
    Description
    For
  • BIBBLR10
    Replacement locking ring for key with bore 10 mm
    Rotaflo® stopcocks
    1 * 5 items - 35,40 €