Tip boxes, dispolab®
Box for refill tips in rack, with transparent snap-on lid, Yellow
Catalog # KART9756-06
Supplier: KARTELL
Jump to:
Product Details & Documents
Empty boxes for use with refill tips.
- Semitransparent boxes in PC have a horizontal sliding cover
- Yellow boxes in PP feature a snap-on cover
Specifications
- Colour:Yellow
- Description:Box for refill tips in rack, with transparent snap-on lid