Cavity slides
Cavity slides with PTFE coating, Black, 3, 14 mm
631-0467
: Epredia
These autoclavable slides are specially designed for immunofluorescence.
- Adhesive surface and hydrophobic mask
- Hydrophobic epoxy- or PTFE-coating
- (*) with rings or wells
Developped for clinical diagnosis applications.
: Available with rings or wells (*).
- No. of wells:3
- Colour:Black
- Description:Cavity slides with PTFE coating
- Seq:018
- Well Ø:14 mm