Orders LinkContactUsLinkComponent
Accessories for analytical Mill, A 11 basic
Accessories for analytical Mill, A 11 basic
Double beater, Use up to Mohs hardness 3. Can only be Used with 250 ml grinding chamber
Catalog # 431-2113
Supplier:   IKA
Accessories for analytical Mill, A 11 basic
Catalog # 431-2113
Supplier:  IKA
Accessories for analytical Mill, A 11 basic
Accessories for analytical Mill, A 11 basic
Double beater, Use up to Mohs hardness 3. Can only be Used with 250 ml grinding chamber
Price
586,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Double beater
  • Seq:
    06
  • For:
    Use up to Mohs hardness 3. Can only be Used with 250 ml grinding chamber