Hook connectors
Hook connector, Steel, powder coated, 13 mm
241-0382
Nickel plated brass screw with plastic handle.
- For rods up to 13 mm
- Angle: 90°
- Description:Hook connector
- Material:Steel, powder coated
- Seq:04
- For rod Ø:13 mm
Nickel plated brass screw with plastic handle.