Microtubes, with Thread
Micro tubes with neutral screw cap enclosed, without skirted base, 1,5 ml, 5000/case
Catalog # SARS72.692
Supplier: SARSTEDT
Jump to:
Product Details & Documents
PP, translucent.
- Safe screw cap with O-ring
- Sterile or non sterile
- Autoclavable
For transport, storage and sample processing in the laboratory.
Specifications
- Packed:5000/case
- Capacity:1,5 ml
- Description:Micro tubes with neutral screw cap enclosed, without skirted base
- Seq:12