Orders LinkContactUsLinkComponent
Gel casting system, MultiCaster
MultiCaster, orange casting stand, UVT tray, 2 reversible combs and a red plate
Catalog # 700-4211
Supplier:   EMBITEC
Gel casting system, MultiCaster
Catalog # 700-4211
Supplier:  EMBITEC
Gel casting system, MultiCaster
Gel casting system, MultiCaster
MultiCaster, orange casting stand, UVT tray, 2 reversible combs and a red plate
Price
278,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Moulded PC. Compatible with the RunOne™ electrophoresis system.
  • Simple to use - no gaskets, clamps or tape required
  • Flexible with options for well size, well configuration, number of wells and resolving area
Specifications
  • Description:
    MultiCaster, orange casting stand, UVT tray, 2 reversible combs and a red plate
  • Seq:
    02
  • Cat. No.:
    700-4211
  • Gel dimensions:
    115×89