Orders LinkContactUsLinkComponent
Adhesion slides
Adhesion slides
Adhesion slides with 12 wells (Ø = 5 mm)
Catalog # 631-9533
Supplier:   MARIENFELD
Adhesion slides
Catalog # 631-9533
Supplier:  MARIENFELD
Adhesion slides
Adhesion slides
Adhesion slides with 12 wells (Ø = 5 mm)
Price
880,87 €
Log in for availability
1 * 50 items
Product Details & Documents
Adhesion slides with frosted ends and 90° ground edges.
Specifications
  • Description:
    Adhesion slides with 12 wells (Ø = 5 mm)