Platinum wire
Platinum wire, Ø 0,8 mm
631-7106
: CHEMPUR
Platinum 99,9%.
Platinum wire is used in many applications including gasket or O-ring fabrication, thermocouples, electrical contacts, loops and heater coils.
- Description:Platinum wire, Ø 0,8 mm
- Seq:06