Orders LinkContactUsLinkComponent
Folding precision magnifiers
Folding precision magnifiers
Folding magnifier, aplanatic, 23 mm
Catalog # 630-8234
Supplier:   ESCHENBACH OPTIK
Folding precision magnifiers
Catalog # 630-8234
Supplier:  ESCHENBACH OPTIK
Folding precision magnifiers
Folding precision magnifiers
Folding magnifier, aplanatic, 23 mm
Price
87,40 €
Log in for availability
1 * 1 items
Product Details & Documents
Swivelling black metallic magnifier with loop, lens frame made from black painted metal.
  • Protective case can serve as a handle
  • Provide distortion-free images
  • Chrome plated metal
Specifications
  • Ø:
    23 mm
  • Magnification:
    12×
  • Description:
    Folding magnifier, aplanatic
  • Seq:
    003