Pipette containers, VARICON
Pipette container for short pipettes, VARICON RK, 60x145-235 mm
Catalog # 612-1850
Supplier: schuett-biotec GmbH
Jump to:
Product Details & Documents
These containers are made of two overlapping aluminium parts, which can be pushed in and out of each other in telescope fashion to match the length/voluminal of the pipettes.
Specifications
- Ø×L:60x145-235 mm
- Description:Pipette container for short pipettes, VARICON RK
- Seq:01
Accessories
Product Information
- Catalog #AvailabilityPrice
- Silicone insert (2 mm thick), prevents damage to pipettes caused by sudden shock, for pipette containers VARICON RS (lower part) and SPECICON (lower and upper part)Silicone insert (2 mm thick), prevents damage to pipettes caused by sudden shock, for pipette containers VARICON RS (lower part) and SPECICON (lower and upper part)SCUT3523213Log in for availability1 * 1 items - 5,90 €