Scalpel handles
No. 1, Standard, 130 mm
Catalog # 233-5201
Supplier: BAYHA
Jump to:
Product Details & Documents
Stainless steel.
- Available in solid form or as version with hollow handle (233-5203)
- Large selection of sterile and non sterile scalpel blades available; all blade shapes fit into any BAYHA handle
- Proven latch system for tight fit of the blade in the handle
- Easy and safe exchange of scalpel blades
Specifications
- Type:No. 1
- Description:Standard
- Length:130 mm