Orders LinkContactUsLinkComponent
Scissors
Scissors
Sharp/sharp, 255 mm
Catalog # 233-1146
Supplier:   WEDO WERNER DORSCH
Scissors
Catalog # 233-1146
Supplier:  WEDO WERNER DORSCH
Scissors
Scissors
Sharp/sharp, 255 mm
Price
11,30 €
Log in for availability
1 * 1 items
Product Details & Documents
General purpose scissors manufactured from stainless steel (polished).
Specifications
  • Type:
    Sharp/sharp
  • Length:
    255 mm
  • Seq:
    06