Orders LinkContactUsLinkComponent
Forceps, ESD-safe
Forceps, ESD-safe
Curved, hook-shaped fine tips, NK6, Black, 120 mm
Catalog # 232-2171
Supplier:   OUTILS RUBIS
Forceps, ESD-safe
Catalog # 232-2171
Supplier:  OUTILS RUBIS
Forceps, ESD-safe
Forceps, ESD-safe
Curved, hook-shaped fine tips, NK6, Black, 120 mm
Price
17,70 €
Log in for availability
1 * 1 items
Product Details & Documents
Conductive Vestamid with 25% fibre glass reinforcement, black. Precise, robust and virtually unbreakable forceps. Very precise; when pressure is applied during work, the tips don’t open.
  • Electrically conductive
  • Very high chemical resistance to acids and bases
  • Heat resistant up to 175 °C
DIN 53482
Certifications: DIN 53482
Specifications
  • Type:
    NK6
  • Colour:
    Black
  • Description:
    Curved, hook-shaped fine tips
  • Length:
    120 mm