Orders LinkContactUsLinkComponent
Scoops
Scoops
85 ml, 155 mm
Catalog # 231-0311
Supplier:   SCHROEDER WILESCO
Scoops
Catalog # 231-0311
Supplier:  SCHROEDER WILESCO
Scoops
Scoops
85 ml, 155 mm
Price
2,30 €
Log in for availability
1 * 1 items
Product Details & Documents
Aluminium, smooth. For collecting or measuring powders, granulates or pastes.
  • Good chemical resistance
  • Excellent thermal resistance
  • Light, robust and easy to use
Specifications
  • Capacity:
    85 ml
  • Length:
    155 mm