Orders LinkContactUsLinkComponent
Scoops
Scoops
2500 ml, 430 mm
Catalog # 231-0309
Supplier:   SCHROEDER WILESCO
Scoops
Catalog # 231-0309
Supplier:  SCHROEDER WILESCO
Scoops
Scoops
2500 ml, 430 mm
Price
32,80 €
Log in for availability
1 * 1 items
Product Details & Documents
Aluminium, smooth. For collecting or measuring powders, granulates or pastes.
  • Good chemical resistance
  • Excellent thermal resistance
  • Light, robust and easy to use
Specifications
  • Capacity:
    2500 ml
  • Length:
    430 mm