Orders LinkContactUsLinkComponent
Scoops
Scoops
240 ml, 250 mm
Catalog # 231-0304
Supplier:   SCHROEDER WILESCO
Scoops
Catalog # 231-0304
Supplier:  SCHROEDER WILESCO
Scoops
Scoops
240 ml, 250 mm
Price
7,50 €
Log in for availability
1 * 1 items
Product Details & Documents
Aluminium, smooth. For collecting or measuring powders, granulates or pastes.
  • Good chemical resistance
  • Excellent thermal resistance
  • Light, robust and easy to use
Specifications
  • Capacity:
    240 ml
  • Length:
    250 mm