Assortment Boxes, PS
Assortment box, with 7 compartments, Red, 210×120×36 mm, PS
216-3716
These assortment boxes made of high impact strength polystyrene (PS) are suitable for packaging purposes and create order in workshops and in household.
- Available either with versatile compartment subdivisions and versions or without compartments
- Transparent lid and bottom for optimum display of the contents or in colours suitable for foods
- Printing possible on request
- Colour:Red
- Description:Assortment box, with 7 compartments
- Material:PS
- Seq:06
- W×D×H:210×120×36 mm