ContactUsLinkComponent
Container multipurpose, 2000ml, with snap-on lid, length 103mm, breadth 255mm, height 94mm, container high density polyethylene, lid low density polyethylene 1 * 1 items
  216-3417
Container multipurpose, 2000ml, with snap-on lid, length 103mm, breadth 255mm, height 94mm, container high density polyethylene, lid low density polyethylene 1 * 1 items
  216-3417
Frequently Bought Together