Orders LinkContactUsLinkComponent
Jars, with twist-off thread
Jars, with twist-off thread
250 ml, 65,0 mm, 122,0 mm, 58 mm
Catalog # 216-3043
Supplier:   MUELLER KREMPEL
Jars, with twist-off thread
Catalog # 216-3043
Supplier:  MUELLER KREMPEL
Jars, with twist-off thread
Jars, with twist-off thread
250 ml, 65,0 mm, 122,0 mm, 58 mm
Price
104,00 €
Log in for availability
1 * 48 items
Product Details & Documents
Amber glass, without lid.
  • Good chemical and physical properties
  • For uses without high thermal load
  • Matching metal lid available as an separately (see 216-3661 and 216-3663)
Well suited for sampling, storage and transport of powders, pastes and liquids. Amber coloured to protect light-sensitive substances.
Specifications
  • Ext.Ø:
    65,0 mm
  • Thread:
    58 mm
  • Capacity:
    250 ml
  • Height:
    122,0 mm
  • Seq:
    04