Orders LinkContactUsLinkComponent
Threaded closures for rectangular bottles
Threaded closures for rectangular bottles
Tamper-proof Screw-caps, White
Catalog # 215-9226
Threaded closures for rectangular bottles
Catalog # 215-9226
Supplier:  
Threaded closures for rectangular bottles
Tamper-proof Screw-caps, White
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
CAP SULES PP SIZE 28MM 1 * 100 items Catalog #215-3237
Product 215-3237 is similar to 215-9226 but manufacturer and specifications may vary.
Product Details & Documents
PP, white.
  • Tamper-evident closure
  • Self-sealing with internal cone
  • Can only be used for rectangular bottles 215-8331 to 215-8334
Specifications
  • Colour:
    White
  • Description:
    Tamper-proof Screw-caps