Orders LinkContactUsLinkComponent
Bottles, narrow neck, rectangular, LDPE, with thread
Bottles, narrow neck, rectangular, LDPE, with thread
125 ml
Catalog # 215-8331
Supplier:   MUELLER KREMPEL
Bottles, narrow neck, rectangular, LDPE, with thread
Catalog # 215-8331
Supplier:  MUELLER KREMPEL
Bottles, narrow neck, rectangular, LDPE, with thread
Bottles, narrow neck, rectangular, LDPE, with thread
125 ml
Price
1,90 €
Log in for availability
1 * 1 items
Product Details & Documents
LDPE, translucent, without screw cap. Ideal for storing and shipping liquids. Graduated.
  • Good chemical resistance
  • Heat resistant up to +80 °C
  • Side ribbing for better grip
 : The bottles are supplied without caps. For suitable caps for 125/500 and 1000 ml please see 215-9226 (for 250 ml botlles see MUEL14.063.01).
Specifications
  • Capacity:
    125 ml