Trolleys, Variocar® 60 basic
Trolley, with three drawers, 600×450×730 mm
Catalog # 139-9974
Supplier: HAEBERLE GmbH + Co. KG
Jump to:
Product Details & Documents
Multipurpose trolleys with chromium plated frame and shelves.
- White shelves and drawers with designed drawer handles in different colours
- Castors of 75 mm Ø each
- Total loading capacity up to 50 kg
Specifications
- Description:Trolley, with three drawers
- Seq:02
- W×D×H:600×450×730 mm