Wet Digester for ecological preincineration with options to automatically remove and neutralize fumes. Main features: up to 6 sample s can be processed in one run, programmable temp 1 * 1 items
Catalog # 458-2201
Supplier: BUCHI
Frequently Bought Together