Orders LinkContactUsLinkComponent
Conical separating funnels
Conical separating funnels
50 ml, 19/26 NS, 2 ml
Catalog # 527-1201
Supplier:   Lenz Laborglas
Conical separating funnels
Catalog # 527-1201
Supplier:  Lenz Laborglas
Conical separating funnels
Conical separating funnels
50 ml, 19/26 NS, 2 ml
Price
58,30 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Both with or without graduations.
Documents
Specifications
  • Socket size:
    19/26 NS
  • Capacity:
    50 ml
  • Division:
    2 ml