Erlenmeyer flasks with screw caps
100 ml, 109 mm, 64 mm, GL 25
Catalog # 214-1206
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with DIN thread and red PBT closure.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and a white labelling area. Screw cap with PTFE coated seal.
Specifications
- Ext.Ø:64 mm
- Thread:GL 25
- Capacity:100 ml
- Height:109 mm