Orders LinkContactUsLinkComponent
Adapters, glass, cone/screwthread
Adapters, glass, cone/screwthread
19/26 NS, GL 18, 7,5 - 9,0 mm
Catalog # 201-2116
Supplier:   Lenz Laborglas
Adapters, glass, cone/screwthread
Catalog # 201-2116
Supplier:  Lenz Laborglas
Adapters, glass, cone/screwthread
Adapters, glass, cone/screwthread
19/26 NS, GL 18, 7,5 - 9,0 mm
Price
23,00 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread adapters, with screw-cap with aperture and silicone sealing ring with PTFE washers.
Documents
Specifications
  • Cone size:
    19/26 NS
  • Thread:
    GL 18
  • Tube ext.Ø:
    7,5 - 9,0 mm
  • Seq:
    06