Orders LinkContactUsLinkComponent
Stopcocks for separating funnels, DURAN®
Stopcocks for separating funnels, DURAN®
12,5 NS
Catalog # SCOT286430807
Supplier:   DWK Life Sciences
Stopcocks for separating funnels, DURAN®
Catalog # SCOT286430807
Supplier:  DWK Life Sciences
Stopcocks for separating funnels, DURAN®
12,5 NS
Price
435,00 €
Log in for availability
1 * 25 items
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, grounded, without key.
  • Very good chemical resistance
  • Very good temperature resistance
With short inlet. For glass manipulators.
Specifications
  • Spigot:
    12,5 NS