Orders LinkContactUsLinkComponent
Electrode Cables
Electrode Cables
BNC connector for pH meters with BNC for example Hach, Orion, Mettler, 1 m
Catalog # 662-6193
Supplier:   SI Analytics
Electrode Cables
Catalog # 662-6193
Supplier:  SI Analytics
Electrode Cables
Electrode Cables
BNC connector for pH meters with BNC for example Hach, Orion, Mettler, 1 m
Price
67,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Electrode cables with a range of connectors to fit most pH meters.
Specifications
  • Description:
    BNC connector for pH meters with BNC for example Hach, Orion, Mettler
  • Length:
    1 m
  • Seq:
    03