Gooseneck light guides
Fibre optic illumination for stereo microscopy. To be used with cold light sources.
In all its microscopy light guides, SCHOTT uses the new environmentally friendly glass optical fibers PURAVIS®. Not just the fibers themselves, but also the entire proprietary manufacturing process are lead free and avoid the use of arsenic and antimony. Due to superior physical and chemical properties, PURAVIS® glass optical fibers provide a high performance, superior longevity, improved transmission in the near UV and outstanding transmission of white light.
Documents
KL Series Fiber Optic Illumination for Stereo Microscopy
Product Literature
SCOC154101
Images 360
TDS Gooseneck Light Guides
Datasheet
- Description:2-arm gooseneck light guide, optic fiber bundle, Ø 3,5 mm, L=500 mm
- Seq:325
- For:KL 300 LED