Orders LinkContactUsLinkComponent
Gooseneck light guides
Gooseneck light guides
1-arm gooseneck light guide, optic fiber bundle, Ø 3,5 mm, L=500 mm, KL 300 LED
Catalog # 631-0001
Gooseneck light guides
Catalog # 631-0001
Gooseneck light guides
Gooseneck light guides
1-arm gooseneck light guide, optic fiber bundle, Ø 3,5 mm, L=500 mm, KL 300 LED
Price
154,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Fibre optic illumination for stereo microscopy. To be used with cold light sources.
In all its microscopy light guides, SCHOTT uses the new environmentally friendly glass optical fibers PURAVIS®. Not just the fibers themselves, but also the entire proprietary manufacturing process are lead free and avoid the use of arsenic and antimony. Due to superior physical and chemical properties, PURAVIS® glass optical fibers provide a high performance, superior longevity, improved transmission in the near UV and outstanding transmission of white light.
Specifications
  • Description:
    1-arm gooseneck light guide, optic fiber bundle, Ø 3,5 mm, L=500 mm
  • Seq:
    320
  • For:
    KL 300 LED
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.