Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PURAVIS - Continuous ring light - inner diameter Ø 58 mm - to connect on KL 1500 HAL / KL 1500 compact / KL 1600 LED / 2500 LCD / KL 2500 LED 1 * 1 items
Catalog # 631-0032
Supplier:
SCHOTT AG Lighting and imaging
Expand image
PURAVIS - Continuous ring light - inner diameter Ø 58 mm - to connect on KL 1500 HAL / KL 1500 compact / KL 1600 LED / 2500 LCD / KL 2500 LED 1 * 1 items
Catalog # 631-0032
Supplier:
SCHOTT AG Lighting and imaging
Related Information
Documentation
Datasheet
TDS Ringlights
Documentation
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals