Orders LinkContactUsLinkComponent
Burette stopcocks, straight
Burette stopcocks, straight
Burettes of 50 ml capacity
Catalog # BRND81939
Supplier:   Brand
Burette stopcocks, straight
Catalog # BRND81939
Supplier:  Brand
Burette stopcocks, straight
Burette stopcocks, straight
Burettes of 50 ml capacity
Price
50,90 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass 5,4, amber.
  • Interchangeable key
  • With glass or PTFE key
  • Key with retention device
Stopcock housing with burette tip and key.
Specifications
  • For:
    Burettes of 50 ml capacity
  • Seq:
    04