Orders LinkContactUsLinkComponent
Cavity slides
Cavity slides, Clear, 3, 15 - 18 mm
Catalog # BRND475565
Supplier:   Brand
Cavity slides
Catalog # BRND475565
Supplier:  Brand
Cavity slides
Cavity slides
Cavity slides, Clear, 3, 15 - 18 mm
Price
274,00 €
Log in for availability
1 * 50 items
Product Details & Documents
Cavity slides in hydrolytic class 3 glass.
  • Ground edges
Specifications
  • No. of wells:
    3
  • Colour:
    Clear
  • Description:
    Cavity slides
  • Seq:
    03
  • Well Ø:
    15 - 18 mm