Slide boxes
White, PS, 50, 200×88×32 mm
Supplier: DELTALAB
Jump to:
Product Details & Documents
Stackable items to gain space on the bench.
- Translucent lid to see if the box is empty or not
- No numbering of the slots
Certificates
Search by Lot NumberSpecifications
- No. of slides:50
- Colour:White
- Material:PS
- Seq:02
- W×D×H:200×88×32 mm