Inoculating loops and needles, disposable
Loop, 10 µl, Blue, 50 loops per sleeve, 40 sleeves per case
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
Flexible polymer.
- Sterilised by irradiation
- Individually wrapped
- Length: 200 mm
Documents
Certificates
Search by Lot NumberSpecifications
- Packed:50 loops per sleeve, 40 sleeves per case
- Capacity:10 µl
- Colour:Blue
- Description:Loop
- Seq:05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.