Orders LinkContactUsLinkComponent
Inoculating loops and needles, disposable
Loop, 10 µl, Blue, Individually packed
  391-2082
Inoculating loops and needles, disposable
  391-2082
Inoculating loops and needles, disposable
Inoculating loops and needles, disposable
Loop, 10 µl, Blue, Individually packed
146,00 €
1 * 600 items
Flexible polymer.
  • Sterilised by irradiation
  • Individually wrapped
  • Length: 200 mm
  • Packed:
    Individually packed
  • Capacity:
    10 µl
  • Colour:
    Blue
  • Description:
    Loop
  • Seq:
    04