Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Cryoboxes with grids 9 x 9 places, PP yellow, format 133 x 133 x 52 mm, for cryo tubes 1,2 - 2 mL (Ø 12,8mm) 1 * 5 items
479-5166
:
RATIOLAB
Cryoboxes with grids 9 x 9 places, PP yellow, format 133 x 133 x 52 mm, for cryo tubes 1,2 - 2 mL (Ø 12,8mm) 1 * 5 items
479-5166
:
RATIOLAB
Frequently Bought Together
VWR®, Universal Filter Tips
: VWR Collection
VWR®, Microtube Storage Boxes
: VWR Collection
CRYOBOX IN PP 81 POS. H50MM NATURAL 1 * 5 items
: RATIOLAB
Pasteur pipettes, glass
: DWK Life Sciences
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Hypodermic needles, HENKE-JECT® for single use
: HENKE SASS WOLF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
FreezerBondz™ Ultra-Thin Cryogenic Polyester Labels for M610, M611, BMP61 Printers
: Brady
OATMEAL AGAR 1 * 250 g
: Merck
Corning® rat collagen I (from tail tendon)
: Corning