SONOPULS HD 2200 - SET, ULTRASONIC HOMOGENIZER, 20 KHZ, 230 V 50/60 HZ, READY-TO-OPERATE BASIC EQUIPMENT FOR VOLUMES FROM 20 ML TO 900 ML, HF-OUTPUT MAX. 200 WEFF. 1 * 1 items
Catalog # 431-5504
Supplier: BANDELIN-ELECTRONIC
Frequently Bought Together