Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Retort Rods
Stand rods
Stand rods
Print
Stand rods
12 mm, 1500 mm
Catalog # 241-7172
Supplier:
USBECK Laborgeräte
Stand rods
Catalog # 241-7172
Supplier:
USBECK Laborgeräte
Expand image
Return to Top
Stand rods
12 mm, 1500 mm
Specifications
Add to cart
Jump to:
Specifications
Price
29,00 €
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Stainless steel.
Robust, very stable
Excellent chemical resistance
Specifications
Ø:
12 mm
Length:
1500 mm
Seq:
16