Orders LinkContactUsLinkComponent
Stand rods
Stand rods
12 mm, 600 mm
Catalog # 241-7161
Supplier:   USBECK Laborgeräte
Stand rods
Catalog # 241-7161
Stand rods
Stand rods
12 mm, 600 mm
Price
7,40 €
Log in for availability
1 * 1 items
Product Details & Documents
Steel, galvanised.
  • Good chemical resistance
  • Very sturdy
Specifications
  • Ø:
    12 mm
  • Length:
    600 mm