Orders LinkContactUsLinkComponent
Stand rods
12 mm, 750 mm
Catalog # 241-7142
Supplier:   USBECK Laborgeräte
Stand rods
Catalog # 241-7142
Stand rods
Stand rods
12 mm, 750 mm
Price
16,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel.
  • Robust, very stable
  • Excellent chemical resistance
Specifications
  • Ø:
    12 mm
  • Length:
    750 mm
  • Seq:
    13