Laboratory scissors, straight
Sharp/blunt, 160 mm
233-1222
: RSG
Stainless steel, INOX 4021.
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Designed for rough handling
- Type:Sharp/blunt
- Length:160 mm