Orders LinkContactUsLinkComponent
Laboratory scissors, straight
Laboratory scissors, straight
Sharp/sharp, 130 mm
Catalog # 233-1211
Supplier:   RSG
Laboratory scissors, straight
Catalog # 233-1211
Supplier:  RSG
Laboratory scissors, straight
Laboratory scissors, straight
Sharp/sharp, 130 mm
Price
9,40 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel, INOX 4021.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Designed for rough handling
Specifications
  • Type:
    Sharp/sharp
  • Length:
    130 mm