Orders LinkContactUsLinkComponent
Double spatulas, micro
Double spatulas, micro
210 mm, 5 mm
Catalog # 231-2263
Supplier:   RSG
Double spatulas, micro
Catalog # 231-2263
Supplier:  RSG
Double spatulas, micro
Double spatulas, micro
210 mm, 5 mm
Price
29,90 €
Log in for availability
1 * 5 items
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
  • Both ends have an angled spoon
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Specifications
  • Length:
    210 mm
  • Blade width:
    5 mm