Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Sample vials in Lab File®, WHEATON®

Sample vials in Lab File®, WHEATON®

4,00 ml, 15×45 mm, Amber, No, 13-425
Catalog # 218-2229
Sample vials in Lab File®, WHEATON®
Catalog # 218-2229
Supplier:  
Sample vials in Lab File®, WHEATON®
Sample vials in Lab File®, WHEATON®

4,00 ml, 15×45 mm, Amber, No, 13-425

Discontinued
Product Details & Documents

For storage of reagents and standards. Excellent for small derivatisation reactions. Wheaton 33® low extractable borosilicate glass is ideal for research studies where stability of glass is of prime importance.

Ordering information: Without cap.

Specifications
  • Cap included:
    No
  • Cap Size:
    13-425
  • Ø×H:
    15×45 mm
  • Capacity:
    4,00 ml
  • Colour:
    Amber
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.