Orders LinkContactUsLinkComponent
Caps for tubes
Caps for tubes
Cap for 11 mm tubes, Natural
Catalog # 217-8930
Supplier:   DELTALAB
Caps for tubes
Catalog # 217-8930
Supplier:  DELTALAB
Caps for tubes
Cap for 11 mm tubes, Natural
Price
139,00 €
Log in for availability
1 * 2.000 items
Product Details & Documents
Polyethylene caps for all types of polypropylene and polystyrene tubes.
Specifications
  • Colour:
    Natural
  • Description:
    Cap for 11 mm tubes
  • Seq:
    03