Orders LinkContactUsLinkComponent
Sample container with screw cap
Sample container with screw cap
60 ml, 38 mm, 65 mm, With white label, Blue, Blue, Bulk
Catalog # 216-4051
Supplier:   DELTALAB
Sample container with screw cap
Catalog # 216-4051
Supplier:  DELTALAB
Sample container with screw cap
Sample container with screw cap
60 ml, 38 mm, 65 mm, With white label, Blue, Blue, Bulk
Price
255,00 €
Log in for availability
1 * 600 items
Product Details & Documents
Container PP, natural or blue, with PE screw cap.
  • With or without label
  • Blue containers are ideal for photosensitive samples
  • Bulk or individually wrapped
To store and transport samples securely.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • Ext.Ø:
    38 mm
  • Packed:
    Bulk
  • Type:
    With white label
  • Capacity:
    60 ml
  • Colour:
    Blue
  • Height:
    65 mm
  • Seq:
    01
  • Cap colour:
    Blue