Orders LinkContactUsLinkComponent
Jars with screw cap
180 ml, 54 mm, 120 mm
  216-3183
 :   VITLAB
Jars with screw cap
  216-3183
 :  VITLAB
Jars with screw cap
Jars with screw cap
180 ml, 54 mm, 120 mm
112,00 €
1 * 10 items
White PP.
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Tightly fitting screw lid
To store and transport samples securely.
 : *The product(s) on this page are outside the scope of the EU Directive 2019/904
  • Ext.Ø:
    54 mm
  • Capacity:
    180 ml
  • Height:
    120 mm