Assortment Boxes, PP
Assortment box, with hinged lid and 3×4 compartments, Anthracite, 175×140×30 mm
216-1268
: SEMADENI
These assortment boxes, made of PP, have a transparent stable hinged lid with lock and tamper-evident seals.
- Robust and strong design
- All models can be stacked together
- Lid is fixed on each tray so that the contents don't mix even if box is turned upside down
- Colour:Anthracite
- Description:Assortment box, with hinged lid and 3×4 compartments
- Seq:02
- W×D×H:175×140×30 mm